Sodium hydroxide 1 M NaOH (1N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:100 °C
:0 °C
:1,04 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,II
:GHS05
- Vapor Pressure:3 mmHg ( 37 °C)
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective