Copper(II) cyclohexanebutyrate Cu 15,8% for atomic absorption spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2218-80-6
:
Formula:C₂₀H₃₄CuO₄
MDL Number:MFCD00036399
Melting Pt:126 °C (dec.)
:402.03
Storage Temperature:Ambient
EINECS:218-723-7