Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Butyl-2-thiourea
1-Butyl-2-thiourea ≥98%
1-Butyl-2-thiourea ≥98%
Irritant
1-Butyl-2-thiourea ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
1-Butyl-2-thiourea ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
1516-32-1
:
Formula:
C₅H₁₂N₂S
MDL Number:
MFCD00022173
:
132.23 g/mol
Melting Pt:
76…79 °C
EINECS:
216-165-9
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL09654
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation