Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Galena
Galena naturally occurring mineral, grains ∼ 1.5-4.8 mm (0.06-0.19 in)
Galena naturally occurring mineral, grains ∼ 1.5-4.8 mm (0.06-0.19 in)
:
Thermo Fisher Scientific
Galena naturally occurring mineral, grains ∼ 1.5-4.8 mm (0.06-0.19 in)
:
Thermo Fisher Scientific
:
1314-87-0
:
12179-39-4
:
̍PbS
:
MFCD00016280
:
239.27 g/mol
:
12,05445
:
Ambient
:
215-246-6
:
3077
:
9,III
:
GHS08
:
GHS07
:
GHS09
msds_ALFA042640
Thermo Scientific Acroseal 25 Years