Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

1,2-Dibromo-1-phenylethane ≥97%

1,2-Dibromo-1-phenylethane ≥97%
Certificates
1,2-Dibromo-1-phenylethane ≥97%
1,2-Dibromo-1-phenylethane ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 93-52-7 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 70-74° Boiling Point: 139-140°/15mm UN No.: 3261 MDL No.: MFCD00000138

Synonyms:  (1,2-Dibromoethyl)benzene
  • CAS Number:93-52-7
  • Formula:C₆H₅CHBrCH₂Br
  • MDL Number:MFCD00000138
  • Molecular Weight:263.96 g/mol
  • Boiling Pt:139…140 °C (15 mmHg)
  • Melting Pt:70…74 °C
  • Density:1,74 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-253-4
  • UN:3261
  • ADR:8,III
Product Family Options

Product Information