Burette clamps
Supplier: BOCHEM
Certificates
About this item
Burette clamps made of zinc plated stainless steel.
- Fingers are covered with silicone coating to protect clamped items
- Single sided or double sided available
Burette clamps made of zinc plated stainless steel.