

2-Propanol:Water solution (50:50)% (v/v) for LC-MS, for flushing LC-MS systems, Burdick & Jackson™
Catalog # BJAALC703-1
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle, GL45
- Pk:6 L
- Seq:5198
Specifications
About this item
:
Boiling Pt:82,3 °C (solvent)
Density:0,9189 g/cm³ (20 °C)
Flash Pt:22,78 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
:67-63-0
MDL Number:MFCD00011674