3-Nitrophenol, crystals
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:554-84-7
:
Formula:C₆H₅NO₃
MDL Number:MFCD00007240
:139.11 g/mol
Merck Index:12,06682
Boiling Pt:265 °C (1013 hPa)
Melting Pt:32…35 °C
Density:1,37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-079-8
UN:1663
ADR:6.1,III
:GHS05
:GHS07