Order Entry
Singapore
ContactUsLinkComponent
Eyewash station, First Eyd
Eyewash station, First Eyd
Catalog # KART2384
Supplier:  KARTELL
undefined
Eyewash station, First Eyd
Catalog # KART2384
Supplier:  KARTELL
Specifications
  • Description:
    Emergency eyewash station, without bottle, Italian version
  • Pk:
    1
Specifications
About this item
Emergency eyewash station moulded in strong high impact PS. The bottle is either included or needs to be ordered seperately. It can be filled with distilled water or other medicated liquid for ophthalmic use.
  • Clearly printed instructions (in English or French) are printed on the support rack
  • Easy to attach to the wall
  • PE bottle with eye cup with snap on cap
  • Bottle is carefully designed to allow a quick and immediate flushing to remove foreign matters and prevent serious eye damage