3-Fluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 372-19-0 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 77°(170°F) Refractive Index: 1.5440 Odour: Recognizable Boiling Point: 186-187° Beilstein Reference: 1305471 UN No.: 2941 MDL No.: MFCD00007758
- CAS Number:372-19-0
- Formula:C₆H₆FN
- MDL Number:MFCD00007758
- Molecular Weight:111.12 g/mol
- Boiling Pt:186…187 °C
- Melting Pt:–2 °C
- Density:1,156
- Flash Pt:77 °C (170 °F)
- Storage Temperature:Ambient
- EINECS:206-747-0
- UN:2941
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 372-19-0 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 77°(170°F) Refractive Index: 1.5440 Odour: Recognizable Boiling Point: 186-187° Beilstein Reference: 1305471 UN No.: 2941 MDL No.: MFCD00007758
- CAS Number:372-19-0
- Formula:C₆H₆FN
- MDL Number:MFCD00007758
- Molecular Weight:111.12 g/mol
- Boiling Pt:186…187 °C
- Melting Pt:–2 °C
- Density:1,156
- Flash Pt:77 °C (170 °F)
- Storage Temperature:Ambient
- EINECS:206-747-0
- UN:2941
- ADR:6.1,III
Documents
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



