
2-Fluorobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:445-29-4
:
Formula:C₇H₅FO₂
MDL Number:MFCD00002405
:140.11 g/mol
Boiling Pt:245 °C (1013 hPa)
Melting Pt:122…125 °C
Density:1,46
Storage Temperature:Ambient
EINECS:207-158-1
:GHS07