
5-Nitrosalicylic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-97-9
:
Formula:C₇H₅NO₅
MDL Number:MFCD00007338
:183.12 g/mol
Merck Index:14,6632
Melting Pt:228…236 °C
Density:1,650
EINECS:202-548-8
:GHS07
Storage Temperature:Ambient