
Sodium benzoate ≥99%, purified
:532-32-1
:
:C₆H₅COONa
:MFCD00012463
:144.11 g/mol
:13,08654
:410…430 °C
:1,44 g/cm³ (20 °C)
:>100 °C
:208-534-8
:01-2119460683-35
:GHS07
:Ambient
- Assay (calculated on anhydrous):Min. 99 %
- Heavy metals (as Pb):Max. 20 ppm
- Loss on drying (100°C):Max. 1.0 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.03 %