
Sodium benzoate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # VWRC103974R
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (calculated on dried substance):Min. 99.5 %
- pH (20°C; 5 %):7 - 9
- Insolubility in water:Max. 50 ppm
- Loss on drying (100°C):Max. 1.0 %
- Total Cl (Chlorine):Max. 0.03 %
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 100 ppm
- Cu (Copper):Max. 1 ppm
- Fe (Iron):Max. 1 ppm
- K (Potassium):Max. 0.05 %
- Mg (Magnesium):Max. 50 ppm
- Pb (Lead):Max. 5 ppm
Specification Test Results
About this item
:
:144.11 g/mol
Merck Index:13,08654
Melting Pt:410…430 °C
Density:1,44 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:208-534-8
REACH:01-2119460683-35
:GHS07
Storage Temperature:Ambient
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463