Torque wrenches, Nalgene®
Torque wrench for torque wrench fittings, 0 - 339 N-m, 3/8"
Catalog # NALG2195-6177
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These wrenches can be used to confirm capping machine settings, on-line inspection and to check torques.
- Ensure leakproof sealing of closures
Ordering information: Order fittings that match closure resin.
Specifications
- Description:Torque wrench for torque wrench fittings
- Tightening torque:0 - 339 N-m
- Use with head style:3/8"