Order Entry
Singapore
ContactUsLinkComponent
Torque wrench fittings for Biotainer® closures, Nalgene®
Torque wrench fittings for Biotainer® closures, Nalgene®
  NALG2595-3038
undefined
Torque wrench fittings for Biotainer® closures, Nalgene®
  NALG2595-3038
  • Description:
    Torque wrench fittings for biotainer closures for closure size 38 - 430
  • Material:
    Epoxy
  • Seq:
    01
  • Pk:
    1
  • Type:
    Drive size: 1/4 in.
Epoxy, specifically designed for Nalgene Biotainer closures.
Proper torquing of Nalgene closures is critical to leakproof sealing.