Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-3-iodobenzene
1-Bromo-3-iodobenzene ≥98%
1-Bromo-3-iodobenzene ≥98%
:
Thermo Fisher Scientific
1-Bromo-3-iodobenzene ≥98%
:
Thermo Fisher Scientific
:
591-18-4
:
:
C₆H₄BrI
:
MFCD00001043
:
282.91 g/mol
:
252 °C
:
–9 °C
:
2,219
:
>110 °C (230 °F)
:
209-703-9
:
GHS07
:
Ambient
msds_ALFAA19596
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years