Order Entry
Singapore
ContactUsLinkComponent
 
Irritant
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Catalog # BAKR4018-04
Supplier:  Avantor
CAS Number:  
undefined
Irritant
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid) free acid ≥99%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Catalog # BAKR4018-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99%
  • Assay (C₈H₁₈N₂O₄S):
    ≥99 %
  • Appearance (fine, white, crystalline powder):
    Passes Test
  • Sodium (USP Id.):
    Passes Test
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:
    ≤0.050
  • Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm:
    ≤0.050
  • pH of 5% Solution at 25°C:
    5.0 - 6.5
  • Ash (sulfated):
    ≤0.2 %
  • Arsenic and Antimony (as As):
    ≤0.050 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • QC Supervisor or equivalent to check supplier's COA to ensure proper chemical form.:
    Passes Test

Specification Test Results

About this item

 :
Merck Index:13,04672
Melting Pt:210…215 °C
Density:1,448 g/cm³ (20 °C)
 :238.31 g/mol
EINECS:230-907-9
 :GHS07
Storage Temperature:Ambient
CAS Number:7365-45-9
Formula:C₈H₁₈N₂O₄S
MDL Number:MFCD00006158