$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
2-Bromo-5-chlorobenzoic acid
2-Bromo-5-chlorobenzoic acid ≥98%
2-Bromo-5-chlorobenzoic acid ≥98%
:
Thermo Fisher Scientific
2-Bromo-5-chlorobenzoic acid ≥98%
:
Thermo Fisher Scientific
:
21739-93-5
:
:
C₇H₄BrClO₂
:
MFCD00013982
:
235.46 g/mol
:
152…156 °C
:
Ambient
:
244-559-0
:
2811
:
6.1,III
:
GHS06
msds_ALFAA10103
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years