1-Bromo-2,6-dichlorobenzene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19393-92-1
:
Formula:C₆H₃BrCl₂
MDL Number:MFCD00000574
:225.9 g/mol
Boiling Pt:241…243 °C
Melting Pt:59…62 °C
Density:2,077 g/cm³ (20 °C)
Flash Pt:>100 °C (212 °F)
EINECS:243-018-6
:GHS07
Storage Temperature:Ambient