

2-Ethyl-1-butene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:760-21-4
:
Formula:C₆H₁₂
MDL Number:MFCD00009341
:84.16 g/mol
Boiling Pt:63…64 °C
Melting Pt:–132 °C
Density:0,689
Flash Pt:–26 °C (–14 °F)
Storage Temperature:Ambient
EINECS:212-078-5
UN:3295
ADR:3,II
:GHS02
:GHS07