Disposable particle filtering respirators, FFP1/FFP2/FFP3
Catalog # KIMB64230
Supplier: KIMBERLY CLARK
Specifications
- Type:M10
- Seq:01
- Pk:20
- With valve:-
- Class:FFP1
Specifications
About this item
Moulded masks made from three-layer filter media. Silicone and latex-free.
- Adjustable comfort head straps increase comfort and ensure secure fit
- Colour coding for easy recognition of performance level
- Nose seal is made from water repellent material for better sealing, reduces fogging on safety glasses
- Tested at 120 mg, increases capacity to operate longer in dusty environments
Certifications: EN 149:2001