Squalane
Glass bottle, 0,25 mL
Catalog # EDQMY0000185
Supplier: EDQM
Jump to:
Product Details & Documents
Synonyms: 2,6,10,15,19,23-Hexamethyltetracosane
- CAS Number:111-01-3
- Formula:C₃₀H₆₂
- MDL Number:MFCD00008953
- Molecular Weight:422.82 g/mol
- Merck Index:12,08923
- Boiling Pt:470 °C (1013 hPa)
- Melting Pt:–35 °C
- Density:0,758 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-825-6
Specifications
Specifications
- Pack type:Glass bottle
- Pk:0,25 mL



