Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 96%
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 96%
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 96%
ALFAL16823.06
:
Thermo Fisher Scientific
:
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 96%
ALFAL16823.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
754-96-1
:
:
C₁₀H₆F₁₆O₂
:
MFCD00192200
:
462.13 g/mol
:
132 °C (4 mmHg)
:
132…134 °C
:
Ambient