

tert-Butyl propiolate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13831-03-3
:
Formula:C₇H₁₀O₂
MDL Number:MFCD00060100
Boiling Pt:52…53 °C (27 mmHg)
Melting Pt:18…20 °C
Density:0,919
Flash Pt:28 °C (82 °F)
UN:3272
ADR:3,III
:126.16 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient