Order Entry
Singapore
Orders LinkContactUsLinkComponent
n-Hexane ≥95% for spectrophotometry

n-Hexane ≥95% for spectrophotometry

Flammable
Health
Irritant
Environment
n-Hexane ≥95% for spectrophotometry
Certificates
n-Hexane ≥95% for spectrophotometry
n-Hexane ≥95% for spectrophotometry

2 Options Available Below

Product Details & Documents

Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:14,4694
  • Boiling Pt:69 °C
  • Melting Pt:–95 °C
  • Density:0,659
  • Flash Pt:–23 °C (–9 °F)
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Product Family Options

Product Information