Order Entry
Singapore
ContactUsLinkComponent
Methanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Health
Flammable
Methanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH230-4
Supplier:  BURDICK JACKSON
   Safer Human and Environmental Health
CAS Number:  
undefined
Health
Flammable
Methanol ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH230-4
Supplier:  BURDICK JACKSON
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Carbonyl Compounds:
    0.001 % max.
  • Color (APHA):
    10 max.
  • GC Analysis:
    99.9 % min.
  • HPLC Gradient @ 254 nm:
    0.005 AU max.
  • Residue:
    3 mg/L max.
  • Titrable Acid:
    0.3 µeq/g max.
  • Titrable Base:
    0.2 µeq/g max.
  • UV Cutoff:
    205 nm max.
  • Water by Karl Fischer Titration:
    0.05 % max.

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64,7 °C
Melting Pt:–98 °C
Density:0,792 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS08
 :GHS02
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595