Order Entry
Singapore
Orders LinkContactUsLinkComponent
Methanol, BAKER ANALYZED® ACS, J.T.Baker®

Methanol, BAKER ANALYZED® ACS, J.T.Baker®

Flammable
Toxic
Health
   Sustainable Options Available
Supplier:   Avantor
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
Methanol, BAKER ANALYZED® ACS, J.T.Baker®

3 Options Available Below

Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64,6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0,7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications for Product Family

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99,8 %
  • Appearance:
    clear
  • Carbonyl Compounds (max. 0.001% each of Acetone, Formaldehyde and Acetaldehyde):
    passes test
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 0,001 %
  • Resistivity (at time of bottling):
    min. 0,1 megaohm-cm
  • Solubility in Water:
    passes test
  • Substances Darkened by H2SO4:
    passes test
  • Substances Reducing KMnO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0,0003
  • Titrable Base (meq/g):
    max. 0,0002
  • Water (H2O):
    max. 0,05 %
  • Trace Impurities (in ppm):
    -
  • Aluminium (Al):
    max. 0,5
  • Barium (Ba):
    max. 0,1
  • Boron (B):
    max. 0,02
  • Cadmium (Cd):
    max. 0,05
  • Calcium (Ca):
    max. 0,5
  • Chromium (Cr):
    max. 0,02
  • Cobalt (Co):
    max. 0,02
  • Copper (Cu):
    max. 0,02
  • Iron (Fe):
    max. 0,1
  • Lead (Pb):
    max. 0,1
  • Magnesium (Mg):
    max. 0,1
  • Manganese (Mn):
    max. 0,02
  • Nickel (Ni):
    max. 0,02
  • Tin (Sn):
    max. 0,1
  • Zinc (Zn):
    max. 0,1

Specifications

Specifications

  • Catalog No:
  • BAKR8045.2500
  • BAKR8045.2500PE
  • BAKR8045.9025
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    BAKR8045.2500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - $153.00
    $153.00
    1 * 2,5 L
       $153.00

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    BAKR8045.2500PE
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - $806.00
    $806.00
    1 * 25 L
       $806.00

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    BAKR8045.9025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health