Ethanol absolute analytical reagent
Catalog # APPCA1613.0500GL
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
About this item
:
:46.07 g/mol
Merck Index:12,03806
Boiling Pt:78,3 °C (1013 hPa)
Melting Pt:–117 °C
Density:0,7895 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
REACH:01-2119457610-43
:GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568