

2-Iodoethanol ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:624-76-0
:
Formula:C₂H₅IO
MDL Number:MFCD00002830
:171.97 g/mol
Boiling Pt:85 °C (25 mmHg)
Density:2,202
Flash Pt:65 °C (149 °F)
Storage Temperature:Refrigerator
EINECS:210-861-6
ADR:6.1,I
:Stabilised with copper
UN:2810
:GHS06
:GHS08