3-Chloro-1-propanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane
CAS Number:627-30-5
:
Formula:C₃H₇ClO
MDL Number:MFCD00002943
Boiling Pt:160…162 °C
Melting Pt:–20 °C
Density:1.131
Molecular Weight:94.54 g/mol
Flash Pt:73 °C (163 °F)
Storage Temperature:Ambient
EINECS:210-992-9
UN:2849
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable: