
2-(2-Chloroethoxy)ethanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:628-89-7
:
Formula:C₄H₉ClO₂
MDL Number:MFCD00002870
:124.57 g/mol
Boiling Pt:185 °C
Density:1,180
Flash Pt:90 °C (194 °F)
Storage Temperature:Ambient
EINECS:211-059-9
UN:3334
ADR:9,
:GHS07