1,1-Dichloro-1-fluoroethane ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # VWRC23370.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity:Max. 0.0002 meq/g
- Colouration:Max. 10 APHA
- Carbonyl compounds (as CH3COCH3):Max. 10 ppm
- Evaporation residue:Max. 5 ppm
- Free chlorine:Max. 1 ppm
- Ionised chlorine:Max. 0.5 ppm
- Water:Max. 50 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.1 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Zn (Zinc):Max. 0.01 ppm
Specification Test Results
About this item
:
:116.95 g/mol
Boiling Pt:31,9 °C (1013 hPa)
Melting Pt:–103,5 °C
Density:1,24 g/cm³ (20 °C)
EINECS:404-080-1
UN:3077
ADR:9,III
CAS Number:1717-00-6
Formula:Cl₂FCCH₃