
tert-Butyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-88-5
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00008807
:116.16 g/mol
Merck Index:14,1537
Boiling Pt:94…96 °C
Melting Pt:–62 °C
Density:0,863
Flash Pt:4 °C (39 °F)
Storage Temperature:Ambient
EINECS:208-760-7
UN:1123
ADR:3,II
:GHS02