Water with 0,1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
:76-05-1
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0,9982 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
Specification Test Results
- Trifluoroacetic Acid (v/v):0.09 - 0.11%
- UV Cutoff:<210 nm
- Residue:< 1 mg/L
- Color (APHA):< 10
- HPLC Gradient @ 215 nm:< 0.020 AU
- HPLC Gradient @ 254 nm:< 0.010 AU
- UV Absorbance @ 230 nm:< 0.150 AU
- UV Absorbance @ 250 nm:< 0.010 AU
- UV Absorbance @ 300 nm:< 0.005 AU
- UV Absorbance @ 400 nm:< 0.005 AU