Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
BJAABB333-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Amines by Ninhydrin Test:10 ppm max.
- Free Acid:Passes test
- GC Analysis:99.8 % min.
- Refractive Index (20ºC):1.5085 - 1.5109
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:50 ppm max.
:
:13,08060
:12,4 %(V)
:–41,55 °C
:0,983 g/cm³ (20 °C)
:20 °C
:Ambient
:203-809-9
:1282
:3,II
:79.1 g/mol
:GHS07
:GHS08
:GHS02
:110-86-1
:C₅H₅N
:MFCD00011732