Petroleum spirit ≥99.0% (by GC) (C5 & C6 Isomers) ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:5,9 %(V)
Melting Pt:–73 °C
Density:0,64 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Acidity:Passes test
- Appearance:Passes test
- Initial Boiling Point:35 ºC min.
- Dry Point:60 ºC max.
- Color (APHA):10 max.
- GC Analysis (total C5 and C6 isomers):99 % min.
- Residue after Evaporation:0.001 % max.
- Water by Karl Fischer Titration:0.02 % max.