Petroleum spirit, 20…70 °C ≥99.0% (C5 & C6 Isomers), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
BJAAGC317-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Electron Capture GC (as lindane):2 ppt max.
- Flame Ionization GC (as nonane):1 ppb max.
- GC Analysis (total C5 and C6 isomers):99 % min.
- Refractive Index (20ºC):1.3600 - 1.3700
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.01 % max.
:
:35…60 °C
:–73 °C
:0,64 g/cm3
:–18 °C
:Ambient
:265-151-9
:1268
:3,II
:GHS07
:GHS08
:GHS02
:GHS09
:64742-49-0
:64742-82-1
:MFCD00081849