Petroleum spirit, 20…70 °C ≥99.0% (C5 & C6 Isomers), Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
:64742-49-0
:64742-82-1
:MFCD00081849
:5,9 %(V)
:–73 °C
:0,64 g/cm³ (20 °C)
:–18 °C
:Ambient
:265-151-9
:1268
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
- Acidity:Passes test
- Assay by GC (total C5 and C6 isomers):99 % min.
- Color (APHA):10 max.
- Dry Point:60 ºC max.
- Initial Boiling Point:35 ºC min.
- Residue after Evaporation:5 mg/L max.
- Water by Karl Fischer Titration:0.02 % max.