


Tetrahydrofuran ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
BJAABB343-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- GC Analysis:99.9 % min.
- Peroxides (as H₂O₂) at time of packaging:2 mg/L max.
- Refractive Index (20ºC):1.4065 - 1.4075
- Water by Karl Fischer Titration:100 ppm max.
:
:72.11 g/mol
:12,09351
:11,8 %(V)
:–108,5 °C
:0,8892 g/cm³ (20 °C)
:–14 °C
:Ambient
:203-726-8
:2056
:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:05-2114615188-48
:GHS02
:GHS08
:GHS07
:109-99-9
:C₄H₈O
:MFCD00005356