

Butyl methacrylate 99% (by GC) stabilised
Catalog # ALFAA19068.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance (colour):Clear, colourless to pale yellow
- Form:Liquid
- Assay (GC):>98.5%
- Refractive Index:1.4225 - 1.4255
- Solution Test:10% solution in diethyl ether - clear
Specification Test Results
About this item
CAS Number:97-88-1
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00009444
Boiling Pt:160…163 °C
Melting Pt:–75 °C
Density:0,894
Flash Pt:49 °C (120 °F)
EINECS:202-615-1
UN:2227
ADR:3,III
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
:142.2 g/mol
:GHS02
:GHS07
Storage Temperature:Refrigerator