Toluene ≥99.8% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
BJAAAH347-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Color (APHA):10 max.
- Water by Karl Fischer Titration:0.03 % max.
- GC Analysis:99.8 % min.
- Refractive Index (20ºC):1.4963 - 1.4971
- Residue:3 mg/L max.
- Substances Darkened by Sulfuric Acid:Passes test
- Sulfur Compounds (as S):0.003 % max.
- UV Absorbance @ 300 nm:0.1 AU max.
- UV Absorbance @ 325 nm:0.025 AU max.
- UV Cutoff:284 nm max.
:
:92.14 g/mol
:13,09607
:7,1 %(V)
:–95 °C
:0,867 g/cm³ (20 °C)
:4 °C
:Ambient
:203-625-9
:1294
:3,II
:05-2114615130-69
:GHS07
:GHS08
:GHS02
:108-88-3
:C₆H₅CH₃
:MFCD00008512