N,N-Dimethylformamide ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Catalog # BJAABB075-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Amines (as dimethylamine):3 ppm max.
- GC Analysis:99.9 % min.
- Residue:5 mg/L max.
- UV Absorbance @ 275 nm:0.3 AU max.
- UV Absorbance @ 300 nm:0.05 AU max.
- UV Absorbance @ 350 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:268 nm max.
- Water by Karl Fischer Titration:150 ppm max.
Specification Test Results
About this item
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:15,2 %(V)
Melting Pt:–60,4 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
:GHS07
:GHS08
:GHS02
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284