

Methanol : Acetic acid (95:5) %, Burdick & Jackson™
Catalog # BJAABJ426-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pk:16 L
- Seq:5254
- Pk Info:4 x 4l
Specifications
About this item
:
Merck Index:13,09754
Boiling Pt:71,8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1,490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07