Methanol ≥99.9%, B&J Brand™ for purge and trap GC analysis, for determination volatile organic compounds, Burdick & Jackson™
BJAA232-235
:
- Pack type:Glass bottle
- Pk:2.820 mL
- Seq:2910
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3280 - 1.3288
- Residue:1 mg/L max.
- Volatile Organic Compounds:Passes test
- Water by Karl Fischer Titration:0.3 % max.
:
:32.04 g/mol
:13,05984
:64,7 °C
:–98 °C
:0,792 g/cm³ (20 °C)
:11 °C
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS08
:GHS02
:67-56-1
:CH₃OH
:MFCD00004595