Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%) ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH212-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Color (APHA):10 max.
- Water-Soluble Titrable Acid:0.3 µeq/g max.
- GC Analysis (n-Hexane):60 % min.
- GC Analysis (total C6 isomers):99.9 % min.
- Residue:3 mg/L max.
- Sulfur Compounds (as S):0.005 % max.
- Thiophene:Passes test
- UV Absorbance @ 225 nm:0.05 AU max.
- UV Cutoff:195 nm max.
- Water by Karl Fisher Titration:0.01 % max.
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:7,7 %(V)
Melting Pt:–95 °C
Density:0,659…0.673 g/cm³ (20 °C)
Flash Pt:–26 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311