



Hexane (mixture of isomers) ≥95% (n-hexane) ≥99.9% (by GC) (n-Hexane > 95%), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
BJAA213-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Color (APHA):10 max.
- Electron Capture GC (as lindane):10 ng/L max.
- Flame Ionization GC (as nonane):5 ppb max.
- GC Analysis (n-Hexane):95 % min.
- GC Analysis (total C6 isomers):99.9 % min.
- Residue:1 mg/L max.
- Thiophene:Passes test
- Water by Karl Fisher Titration:0.01 % max.
:
:86.18 g/mol
:13,04712
:68,7 °C
:–95 °C
:0,659…0.673 g/cm³ (20 °C)
:–26 °C
:Ambient
:203-777-6
:1208
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD02179311