



Hexane (mixture of isomers) ≥60,0% (n-hexane) ≥99.9% (by GC), B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:68,7 °C
Melting Pt:–95 °C
Density:0,659…0.673 g/cm³ (20 °C)
Flash Pt:–26 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- GC Analysis (n-Hexane):60 % min.
- GC Analysis (total C6 isomers):99.9 % min.
- Residue:1 mg/L max.
- Water by Karl Fisher Titration:0.01 % max.