

Diethyl ether ≥99.9% (excluding ethanol) stabilised ACS, meets analytical specification of USP for HPLC, for spectrophotometry
BJAAAH106-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Carbonyl (as HCHO):0.001 % max.
- Color (APHA):10 max.
- GC Analysis (excluding ethanol):99.9 % min.
- Peroxides (as H₂O₂) at time of packaging:1 ppm max.
- Refractive Index (20ºC):1.3523 - 1.3533
- Residue:3 mg/L max.
- Titrable Acid:0.2 µeq/g max.
- UV Cutoff:215 nm max.
- Water by Karl Fischer Titration:0.03 % max.
:
:74.12 g/mol
:12,03852
:35 °C
:–117,4 °C
:0,713 g/cm³ (20 °C)
:–45 °C
:Ambient
:200-467-2
:1155
:3,I
:Stabilised with ethanol 2%
:01-2119535785-29
:GHS07
:GHS02
:60-29-7
:(CH₃CH₂)₂O
:MFCD00011646