Order Entry
Singapore
ContactUsLinkComponent
Acetic acid 0,1%
Acetic acid 0,1%
Catalog # BJAA9150
Supplier:  BURDICK JACKSON
CAS Number:  
undefined
Acetic acid 0,1%
Catalog # BJAA9150
Supplier:  BURDICK JACKSON
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152